Bis[3-(triethoxysilyl)propyl]tetrasulfide (∼22,3% S) ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42410-06
42410-14
42410-22
42410-36
CAAA42410-06EA
64.15
CAD
CAAA42410-06
CAAA42410-14
CAAA42410-22
CAAA42410-36
Bis[3-(triethoxysilyl)propyl]tetrasulfide (∼22,3% S) ≥98%
Bis[3-(triethoxysilyl)propyl]tetrasulfide
Applications: Coupling/cross-linking agent for precious metals
CAS No.: 40372-72-3
Molecular Formula: ((CH3CH2O)3SiCH2CH2CH2SS)2
Formula Weight: 538.94
Flash Point: 91°(196°F)
Physical Form: Liquid
Density: 1.074
Boiling Point: 250° dec.
MDL No.: MFCD00053751
Applications: Coupling/cross-linking agent for precious metals
Formula:
C₁₈H₄₂O₆S₄Si₂ Boiling Pt: 250 °C (dec.) Density: 1,074 Flash Pt: 91 °C (196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053751 CAS Number: 40372-72-3 EINECS: 254-896-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...