Poly(2-ethyl-2-oxazoline), M.W. ~500,000

Supplier: Thermo Scientific Chemicals

42398-22 42398-36 42398-A4
CAAA42398-22EA 58.62 CAD
CAAA42398-22 CAAA42398-36 CAAA42398-A4
Poly(2-ethyl-2-oxazoline), M.W. ~500,000
Poly(2-ethyl-2-oxazoline), M.W. ~200,000

Nominal M.W. 500,000


CAS No.: 25805-17-8 Molecular Formula: [-N(COC2H5)CH2CH2-]n Physical Form: Solid MDL No.: MFCD00134377

Formula: (C₅H₉NO)x
Storage Temperature: Ambient
CAS Number: 25805-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR