Poly(2-ethyl-2-oxazoline), M.W. ~50,000
Supplier: Thermo Scientific Chemicals
42396-22
42396-36
42396-A4
CAAA42396-22EA
59.72
CAD
CAAA42396-22
CAAA42396-36
CAAA42396-A4
Poly(2-ethyl-2-oxazoline), M.W. ~50,000
Poly(2-ethyl-2-oxazoline), M.W. ~200,000
Nominal M.W. 50,000
CAS No.: 25805-17-8
Molecular Formula: [-N(COC2H5)CH2CH2-]n
Physical Form: Solid
MDL No.: MFCD00134377
Formula:
(C₅H₉NO)x Storage Temperature: Ambient |
CAS Number:
25805-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...