Sodium deuteroxide 40% (w/w) in D₂O (99.5% D) (w/w)

Supplier: Thermo Scientific Chemicals
Danger

42346-09 42346-18
CAAA42346-09EA 84.21 CAD
CAAA42346-09 CAAA42346-18
Sodium deuteroxide 40% (w/w) in D₂O (99.5% D) (w/w)
Sodium deuteroxide

Packaged in polyethylene bottles


Purity: 99.5% CAS No.: 14014-06-3 Molecular Formula: NaOD Formula Weight: 41.01 Physical Form: Liquid Density: 1.516 Refractive Index: 1.4180 Sensitivity: Air and Moisture Sensitive Appearance: Colorless UN No.: 1824 MDL No.: MFCD00037669

Formula: NaOD
Density: 1.516
Storage Temperature: Ambient
MDL Number: MFCD00037669
CAS Number: 14014-06-3
EINECS: 237-825-2
UN: 1824
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR