Tricosachromium hexacarbide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chromium carbide

42313-09 42313-18 42313-30
CAAA42313-09EA 53.25 CAD
CAAA42313-09 CAAA42313-18 CAAA42313-30
Tricosachromium hexacarbide ≥99.5% (metals basis)
Tricosachromium hexacarbide

Purity: 99.5% CAS No.: 12105-81-6 Molecular Formula: Cr23C6 Formula Weight: 1267.97 Physical Form: -325 Mesh Powder UN No.: 3178 MDL No.: MFCD00678358

Formula: C₆Cr₂₃
MW: 1267.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00678358
CAS Number: 12105-81-6
EINECS: 235-160-2
UN: 3178
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR