Gold plating solution ∼3.7 g/l metal content, electroless
Supplier: Thermo Scientific Chemicals
|
Danger
|
42307-AK
42307-K2
CAAA42307-AKEA
447.14
CAD
CAAA42307-AK
CAAA42307-K2
Gold plating solution ∼3.7 g/l metal content, electroless
Gold plating solution
Applications: For coating nickel or copper
Physical Form: Liquid
Appearance: Colorless to pale yellow
UN No.: 2922
MDL No.: MFCD00003436
Applications: For coating nickel or copper
Storage Temperature:
Ambient |
MDL Number:
MFCD00003436 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...