Tri(1-naphthyl)phosphine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42304-02
42304-04
42304-09
42304-18
CAAA42304-02EA
56.72
CAD
CAAA42304-02
CAAA42304-04
CAAA42304-09
CAAA42304-18
Tri(1-naphthyl)phosphine 97%
Tri(1-naphthyl)phosphine
CAS No.: 3411-48-1
Molecular Formula: (C10H7)3P
Formula Weight: 412.47
Physical Form: Powder
Melting Point: 265-268°
MDL No.: MFCD00046400
Formula:
C₃₀H₂₁P Melting Pt: 265…268 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046400 CAS Number: 3411-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...