Amberlite® IRA-67, Amberlite™ ion exchange resin

Supplier: Thermo Scientific Chemicals

42253-30 42253-A1 42253-A7
CAAA42253-30EA 62.57 CAD
CAAA42253-30 CAAA42253-A1 CAAA42253-A7
Amberlite® IRA-67, Amberlite™ ion exchange resin
Amberlite® IRA-67

Acrylic-DVB; Weakly basic gel type resin


CAS No.: 80747-90-6 Odor: Amine-like MDL No.: MFCD00145567

Storage Temperature: Ambient
MDL Number: MFCD00145567
CAS Number: 80747-90-6
Merck Index: 14,382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR