Amberlite® IRA-200C (Na), Amberlite™ ion exchange resin
Supplier: Thermo Scientific Chemicals
42252-30 42252-A1 42252-A7
CAAA42252-30EA 45.82 CAD
CAAA42252-30 CAAA42252-A1 CAAA42252-A7
Amberlite® IRA-200C (Na), Amberlite™ ion exchange resin
Amberlite® IRA-200C (Na)
Styrene-DVB; Strongly acidic macroreticular cation exchange resin
Odor: Odorless MDL No.: MFCD01864001