Polyethylene oxide, M.W. 300,000
Supplier: Thermo Scientific Chemicals
Synonyms:
PEO
42237-14
42237-22
42237-36
CAAA42237-14EA
45.19
CAD
CAAA42237-14
CAAA42237-22
CAAA42237-36
Polyethylene oxide, M.W. 300,000
Polyethylene oxide
Tg=-67°
CAS No.: 25322-68-3
Molecular Formula: (-CH2CH2O-)n
Physical Form: Powder
Density: 1.21
Refractive Index: 1.4539
Appearance: White to off-white
Odor: Odorless
pH: 5-7
Melting Point: 65°
MDL No.: MFCD00147979
Formula:
(-CH₂CH₂O-)n Melting Pt: 65 °C Density: 1,21 Storage Temperature: Ambient |
MDL Number:
MFCD00147970 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...