Hexavalent Chromium, standard solution, Specpure®, Cr+6 1000 µg/ml

Supplier: Thermo Scientific Chemicals

42234-AE 42234-AP
CAAA42234-AEEA 157.68 CAD
CAAA42234-AE CAAA42234-AP
Hexavalent Chromium, standard solution, Specpure®, Cr+6 1000 µg/ml
Standards Cationic Standards Chromium Standards

Molecular Formula: (NH4)2Cr2O7 in H2O
Physical Form: Liquid
Appearance: Yellow
Odor: Odorless
MDL No.: MFCD00717544

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR