Indium(III) acetate ≥99.99%

Supplier: Thermo Scientific Chemicals

42230-09 42230-18
CAAA42230-09EA 161.16 CAD
CAAA42230-09 CAAA42230-18
Indium(III) acetate ≥99.99%
Indium(III) acetate

Purity: 99.99% CAS No.: 25114-58-3 Molecular Formula: In(OOCCH3)3 Formula Weight: 291.96 Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD00054347

Formula: C₂H₄InO₂
MW: 174.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054347
CAS Number: 25114-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR