(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42120-04
42120-02
CAAAA42120-04EA
276.5
CAD
CAAAA42120-04
CAAAA42120-09
CAAA42120-02
CAAA42120-MC
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Chiral catalyst ligand
Purity: 98%
CAS No.: 100165-88-6
Molecular Formula: C48H40P2
Formula Weight: 678.79
Physical Form: Powder
Melting Point: 255-257°
MDL No.: MFCD00269856
Chiral catalyst ligand
Formula:
C₄₈H₄₀P₂ Melting Pt: 255…257 °C Storage Temperature: Ambient |
MDL Number:
MFCD01311709 CAS Number: 100165-88-6 |
Specification Test Results
Form | Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...