(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthalene, (R)-(+)-TolBINAP
42119-04
42119-02
42119-MC
CAAAA42119-04EA
360.24
CAD
CAAAA42119-04
CAAA42119-02
CAAA42119-MC
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl 98%
(R)-(+)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl
Chiral catalyst ligand
Purity: 98%
CAS No.: 99646-28-3
Molecular Formula: C48H40P2
Formula Weight: 678.79
Physical Form: Powder
Appearance: White
Melting Point: 255-257°
Optical Rotation: +156°(c=0.5 in benzene)
MDL No.: MFCD00269856
Chiral catalyst ligand
Formula:
C₄₈H₄₀P₂ MW: 678.78 g/mol Melting Pt: 255…257 °C Storage Temperature: Ambient |
MDL Number:
MFCD01311709 CAS Number: 99646-28-3 |
Specification Test Results
Form | Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...