Indium(III) isopropoxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
42098-02
42098-04
42098-09
CAAA42098-02EA
87.53
CAD
CAAA42098-02
CAAA42098-04
CAAA42098-09
Indium(III) isopropoxide ≥99.9% (metals basis)
Indium (III) isopropoxide
Precursor used in the preparation of indium oxide containing films
Purity: 99.9%
CAS No.: 38218-24-5
Molecular Formula: C9H21InO3
Formula Weight: 292.08
Physical Form: Powder, Packaged under argon
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 145° dec.
UN No.: 3181
MDL No.: MFCD00210318
Precursor used in the preparation of indium oxide containing films
Formula:
C₉H₂₁InO₃ MW: 292.08 g/mol Melting Pt: 145 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00210318 CAS Number: 38218-24-5 UN: 3181 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...