Cresyl diphenyl phosphate (mixture of analogue) mixture of isomers ≥94%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diphenyl p-tolyl phosphate, Diphenyl Cresyl Phosphate, Phosphoric Acid Cresyl Diphenyl Ester

42092-AE 42092-AP
CAAA42092-AEEA 66.36 CAD
CAAA42092-AE CAAA42092-AP
Cresyl diphenyl phosphate (mixture of analogue) mixture of isomers ≥94%
Cresyl diphenyl phosphate (mixture of analogue)

Purity: 94% CAS No.: 26444-49-5 Molecular Formula: (CH3C6H4O)xP(O)(OC6H5)y Flash Point: 232°(450°F) Physical Form: Liquid Density: 1.21 Refractive Index: 1.5630 Appearance: Pale yellow Melting Point: -38° Boiling Point: 390° UN No.: 3082 MDL No.: MFCD00014920

Formula: C₁₉H₁₇O₄P
MW: 340.3097 g/mol
Boiling Pt: 390 °C
Melting Pt: –38 °C
Density: 1.21
Flash Pt: 232 °C (450 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014920
CAS Number: 26444-49-5
EINECS: 247-693-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR