Neodymium nitride ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

42091-04 42091-09
CAAA42091-04EA 120.24 CAD
CAAA42091-04 CAAA42091-09
Neodymium nitride ≥99.9% (REO, rare earth oxide basis)
Neodymium nitride

Purity: 99.9% CAS No.: 25764-11-8 Molecular Formula: NdN Formula Weight: 158.25 Physical Form: -60 Mesh Powder

Formula: NNd
Storage Temperature: Ambient
MDL Number: MFCD00049990
CAS Number: 25764-11-8
EINECS: 247-246-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR