Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:12.5 wt%), spheres, Ø 4 mm (0.16 in) Type 316
Supplier: Thermo Scientific Chemicals
42021-KM
42021-KN
CAAA42021-KMEA
56.88
CAD
CAAA42021-KM
CAAA42021-KN
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:12.5 wt%), spheres, Ø 4 mm (0.16 in) Type 316
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:12.5 wt%
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...