Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Ø 1 mm (0.04 in) Type 316
Supplier: Thermo Scientific Chemicals
42018-KM
42018-KN
CAAA42018-KMEA
60.83
CAD
CAAA42018-KM
CAAA42018-KN
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), spheres, Ø 1 mm (0.04 in) Type 316
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...