Eicosanedioic acid
Supplier: Thermo Scientific Chemicals
42004-04
42004-09
42004-18
42004-30
CAAA42004-04EA
70.15
CAD
CAAA42004-04
CAAA42004-09
CAAA42004-18
CAAA42004-30
Eicosanedioic acid
Eicosanedioic acid
CAS No.: 2424-92-2
Molecular Formula: C20H38O4
Formula Weight: 342.52
Physical Form: Powder
Melting Point: 127°
MDL No.: MFCD00059627
Formula:
C₂₀H₃₈O₄ Melting Pt: 127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00059627 CAS Number: 2424-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...