Aluminum acrylate
Supplier: Thermo Scientific Chemicals
|
Warning
|
42003-06
42003-14
42003-22
CAAA42003-06EA
99.22
CAD
CAAA42003-06
CAAA42003-14
CAAA42003-22
Aluminum acrylate
Aluminum acrylate
CAS No.: 15743-20-1
Molecular Formula: Al(H2C=CHCOO)3
Formula Weight: 243.17
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
MDL No.: MFCD00078421
Formula:
Al(H₂C=CHCOO)₃ Storage Temperature: Ambient |
MDL Number:
MFCD00078421 CAS Number: 15743-20-1 EINECS: 239-836-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...