Magnesium acrylate
Supplier: Thermo Scientific Chemicals
|
Warning
|
42002-06
42002-14
42002-22
CAAA42002-06EA
59.72
CAD
CAAA42002-06
CAAA42002-14
CAAA42002-22
Magnesium acrylate
Magnesium acrylate
CAS No.: 5698-98-6
Molecular Formula: C6H6MgO4
Formula Weight: 166.42
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Melting Point:
Formula:
C₆H₆MgO₄ Melting Pt: >220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00068285 CAS Number: 5698-98-6 EINECS: 227-177-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...