Yttrium(III) phosphate hydrate ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

41991-04 41991-09
CAAA41991-04EA 88.16 CAD
CAAA41991-04 CAAA41991-09
Yttrium(III) phosphate hydrate ≥99.99% (REO, rare earth oxide basis)
Yttrium(III) phosphate hydrate

Purity: 99.99% CAS No.: 34054-55-2 Molecular Formula: YPO4.xH2O Formula Weight: 183.88(anhy) Physical Form: Powder MDL No.: MFCD00049782

Formula: YPO₄·xH₂O
MW: 183.88 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00049782
CAS Number: 34054-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR