Lanthanum(III) phosphate hydrate ≥99.99%

Supplier: Thermo Scientific Chemicals

41990-06 41990-14
CAAA41990-06EA 82.48 CAD
CAAA41990-06 CAAA41990-14
Lanthanum(III) phosphate hydrate ≥99.99%
Lanthanum(III) phosphate hydrate

Purity: 99.99% CAS No.: 14913-14-5 Molecular Formula: LaPO4.xH2O Formula Weight: 233.88(anhy) Physical Form: Lump Appearance: White to off-white Odor: Odorless MDL No.: MFCD00150248

Formula: LaPO₄.xH₂O
MW: 233.877 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150248
CAS Number: 14913-14-5
EINECS: 237-419-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR