Europium(III) phosphate hydrate ≥99.9%
Supplier: Thermo Scientific Chemicals
|
Warning
|
41989-03
41989-06
CAAA41989-03EA
86.11
CAD
CAAA41989-03
CAAA41989-06
Europium(III) phosphate hydrate ≥99.9%
Europium(III) phosphate hydrate
Purity: 99.9%
CAS No.: 14913-20-3
Molecular Formula: EuO4P.xH2O
Formula Weight: 246.93(anhy)
Physical Form: Chunks/powder
Sensitivity: Hygroscopic
MDL No.: MFCD05664261
Formula:
EuO₄P.xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD05664261 CAS Number: 14913-20-3 EINECS: 236-901-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...