Lead(II) tetrafluoroborate 50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

41956-A1 41956-D5
CAAA41956-A1EA 339.7 CAD
CAAA41956-A1 CAAA41956-D5
Lead(II) tetrafluoroborate 50% (w/w) in aqueous solution
Lead (II) tetrafluoroborate

Applications: tin-lead electroplating


CAS No.: 13814-96-5 Molecular Formula: B2F8Pb Formula Weight: 380.81 Physical Form: Liquid Density: 1.62 Appearance: Colorless UN No.: 2922 MDL No.: MFCD00016270


Used in tin-lead electroplating

Formula: B₂F₈Pb
Density: 1.62
Storage Temperature: Ambient
MDL Number: MFCD00016270
CAS Number: 13814-96-5
EINECS: 237-486-0
UN: 2922
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR