Magnesium citrate dibasic hydrate

Supplier: Thermo Scientific Chemicals

41954-30 41954-A1
CAAA41954-30EA 170.64 CAD
CAAA41954-30 CAAA41954-A1 CAAA41954-A7
Magnesium citrate dibasic hydrate
Magnesium citrate dibasic hydrate

CAS No.: 119851-23-9 Molecular Formula: C6H6MgO7.xH2O Formula Weight: 214.14(anhy) Physical Form: Powder Appearance: White to pale yellow MDL No.: MFCD00151706

Formula: C₆H₆MgO₇·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00151706
CAS Number: 119851-23-9
EINECS: 231-923-9
Merck Index: 14,5663

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR