3-(Benzylamino)propionitrile 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Benzylamino)propanenitrile

41949-14 41949-22
CAAA41949-14EA 140.3 CAD
CAAA41949-14 CAAA41949-22
3-(Benzylamino)propionitrile 97+%
3-(Benzylamino)propionitrile

Purity: 97+% CAS No.: 706-03-6 Molecular Formula: C6H5CH2NHCH2CH2CN Formula Weight: 160.22 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.024 Refractive Index: 1.5308 UN No.: 3276 MDL No.: MFCD00001955

Formula: C₁₀H₁₂N₂
MW: 160.22 g/mol
Density: 1.024
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001955
CAS Number: 706-03-6
EINECS: 211-890-7
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR