Magnesium nitride ≥99.6% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimagnesium dinitride
41946-09
41946-18
CAAA41946-09EA
155.16
CAD
CAAA41946-09
CAAA41946-18
Magnesium nitride ≥99.6% (metals basis)
Magnesium nitride
Purity: 99.6%
CAS No.: 12057-71-5
Molecular Formula: Mg3N2
Formula Weight: 100.95
Physical Form: -325 Mesh Powder
Density: 2.71
Melting Point: 800° dec.
UN No.: 3178
MDL No.: MFCD00016205
Formula:
Mg₃N₂ MW: 100.93 g/mol Melting Pt: 800 °C (dec.) Density: 2.71 Storage Temperature: Ambient |
MDL Number:
MFCD00016205 CAS Number: 12057-71-5 EINECS: 235-022-1 UN: 3178 ADR: 4.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...