Strontium tungsten oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

41899-14 41899-22
CAAA41899-14EA 170.64 CAD
CAAA41899-14 CAAA41899-22
Strontium tungsten oxide ≥99.9% (metals basis)
Strontium tungsten oxide

Purity: 99.9% CAS No.: 13451-05-3 Molecular Formula: O4SrW Formula Weight: 335.46 Physical Form: -200 Mesh Powder Density: 6.187 Appearance: White Odor: Odorless MDL No.: MFCD00049562

Formula: O₄SrW
Density: 6,187
Storage Temperature: Ambient
MDL Number: MFCD00049562
CAS Number: 13451-05-3
EINECS: 236-617-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR