Lithiumbis(trimethylsilyl)amide 0.9-1.1 M in hexane
Supplier: Thermo Scientific Chemicals
|
Danger
|
Packaged under argon in resealable ChemSeal™ bottles.
41872-22
41872-36
CAAA41872-22EA
126.08
CAD
CAAA41872-22
CAAA41872-36
Lithiumbis(trimethylsilyl)amide 0.9-1.1 M in hexane
Lithium bis(trimethylsilyl)amide
CAS No.: 4039-32-1
Molecular Formula: C6H18LiNSi2
Formula Weight: 167.33
Flash Point: -31°(-24°F)
Physical Form: Liquid
Density: 0.716
Appearance: Colorless to pale yellow
Beilstein Reference: 8136861
UN No.: 2924
MDL No.: MFCD00008261
Formula:
C₆H₁₈LiNSi₂ Density: 0.716 Flash Pt: –31 °C (–24 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008261 CAS Number: 4039-32-1 EINECS: 223-725-6 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...