Copper(II) hexafluoroacetylacetonate hydrate ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
41870-04
41870-09
CAAA41870-02EA
47.38
CAD
CAAA41870-02
CAAA41870-04
CAAA41870-09
Copper(II) hexafluoroacetylacetonate hydrate ≥99.99% (metals basis)
Copper (II) hexafluoroacetylacetonate hydrate
Purity: 99.99%
CAS No.: 155640-85-0
Molecular Formula: Cu(CF3COCHCOCF3)2.xH2O
Formula Weight: 477.64(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Boiling Point: 220° dec.
MDL No.: MFCD00151019
Formula:
C₁₀H₂CuF₁₂O₄·xH₂O Boiling Pt: 220 °C (dec.) Melting Pt: ∼ 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151019 CAS Number: 155640-85-0 EINECS: 238-850-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...