Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')iron(III) ≥99.9% (metals basis), crystals, recrystallized
Supplier: Thermo Scientific Chemicals
|
Warning
|
41862-03
41862-06
41862-14
CAAA41862-03EA
88.95
CAD
CAAA41862-03
CAAA41862-06
CAAA41862-14
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')iron(III) ≥99.9% (metals basis), crystals, recrystallized
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')iron(III)
Purity: 99.9%
CAS No.: 14876-47-2
Molecular Formula: C33H57FeO6
Formula Weight: 605.66
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Bright red
Odor: Odorless
Melting Point: 148°
Boiling Point: 300° dec.
MDL No.: MFCD00067465
Formula:
C₃₃H₅₇FeO₆ Boiling Pt: 300 °C (dec.) Melting Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067465 CAS Number: 14876-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...