Bismuth tungsten oxide 99,9

Supplier: Thermo Scientific Chemicals

41854-14 41854-22
CAAA41854-14EA 208.56 CAD
CAAA41854-14 CAAA41854-22
Bismuth tungsten oxide 99,9
Bismuth tungsten oxide

Purity: 99.9% CAS No.: 13595-87-4 Molecular Formula: Bi2(WO4)3 Formula Weight: 1161.48 Physical Form: -200 Mesh Powder Odor: Odorless MDL No.: MFCD00046190

Formula: Bi₂(WO₄)₃
MW: 1161.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046190
CAS Number: 13595-87-4
EINECS: 237-033-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR