Molybdenum(V) isopropoxide 5% (w/v) in isopropanol (IPA) , 99.6% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Packaged under argon in resealable ChemSeal™ bottles.
41836-AC
41836-AE
CAAA41836-ACEA
240.16
CAD
CAAA41836-AC
CAAA41836-AE
Molybdenum(V) isopropoxide 5% (w/v) in isopropanol (IPA) , 99.6% (metals basis)
Molybdenum(V) isopropoxide
Packaged under Argon in resealable ChemSeal™ bottles
Purity: 99.6%
CAS No.: 209733-38-0
Molecular Formula: C15H35MoO5
Formula Weight: 391.04
Flash Point: 12°(54°F)
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Yellow to brown
Odor: Alcohol-like
UN No.: 1219
MDL No.: MFCD00210636
Formula:
C₁₅H₃₅MoO₅ MW: 391.39 g/mol Flash Pt: 12 °C (54 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00210636 CAS Number: 209733-38-0 UN: 1219 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...