Nickel(II) wolframate ≥99.9% (metals basis excluding Co) (max. 100 ppm Co)

Supplier: Thermo Scientific Chemicals
Danger

41826-14 41826-22
CAAA41826-14EA 278.08 CAD
CAAA41826-14 CAAA41826-22
Nickel(II) wolframate ≥99.9% (metals basis excluding Co) (max. 100 ppm Co)
Nickel(II) wolframate

Purity: 99.9% CAS No.: 14177-51-6 Molecular Formula: NiWO4 Formula Weight: 306.53 Physical Form: -100 Mesh Powder Odor: Odorless MDL No.: MFCD00016268

Formula: NiO₄W
Storage Temperature: Ambient
MDL Number: MFCD00016268
CAS Number: 14177-51-6
EINECS: 238-032-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR