Palladium 5% on alumina, pellets 3 mm
Supplier: Thermo Scientific Chemicals
41825-06
41825-14
41825-22
CAAA41825-06EA
130.67
CAD
CAAA41825-06
CAAA41825-14
CAAA41825-22
Palladium 5% on alumina, pellets 3 mm
Palladium
Applications: Hydrogen removal "getter"
CAS No.: 7440-05-3
Odor: Odorless
MDL No.: MFCD03613602
Applications: Hydrogen removal "getter"
Formula:
Pd MW: 106.420 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...