Poly(styrene-divinylbenzene), cross-linked with 1% DVB 200-400 mesh

Supplier: Thermo Scientific Chemicals

41800-06 41800-14 41800-22
CAAA41800-06EA 48.51 CAD
CAAA41800-06 CAAA41800-14 CAAA41800-22
Poly(styrene-divinylbenzene), cross-linked with 1% DVB 200-400 mesh
Poly(styrene-divinylbenzene)

For the preparation of functionalized beads used in solid-phase synthesis


CAS No.: 9003-70-7 Physical Form: Beads Odor: Odorless MDL No.: MFCD00146442


Applications: For the preparation of functionalized beads used in solid-phase synthesis

Formula: [-CH₂CH(C₆H₅)-]x[-CH₂CH[C₆H₄(CHCH₂)]-]y
Storage Temperature: Ambient
MDL Number: MFCD00146442
CAS Number: 9003-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR