Humic acid ≥98%

Supplier: Thermo Scientific Chemicals

41747-06 41747-14
CAAA41747-06EA 71.57 CAD
CAAA41747-06 CAAA41747-14
Humic acid ≥98%
Humic acid

Applications: Metal chelator; pigments
Slightly soluble in water, with much swelling


CAS No.: 1415-93-6 Physical Form: Crystalline powder Appearance: Dark brown Solubility: Slightly soluble in water, with much swelling MDL No.: MFCD00147177


Applications: Metal chelator; pigments

Formula: C₉H₉NO₆
Storage Temperature: Ambient
MDL Number: MFCD00147177
CAS Number: 1415-93-6
EINECS: 215-809-6
Merck Index: 14,4752

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR