Tetrabutylphosphonium hydroxide 40% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
41718-AD
41718-AK
41718-K2
CAAA41718-ADEA
90.38
CAD
CAAA41718-AD
CAAA41718-AK
CAAA41718-K2
Tetrabutylphosphonium hydroxide 40% (w/w) in aqueous solution
Tetrabutylphosphonium hydroxide
CAS No.: 14518-69-5
Molecular Formula: C16H37OP
Formula Weight: 276.45
Physical Form: Liquid
Density: 0.989
Refractive Index: 1.4120
UN No.: 3267
MDL No.: MFCD00068456
Formula:
C₁₆H₃₇OP MW: 276.45 g/mol Density: 0.989 Storage Temperature: Ambient |
MDL Number:
MFCD00068456 CAS Number: 14518-69-5 EINECS: 238-528-0 UN: 3267 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...