Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate

Supplier: Thermo Scientific Chemicals
Warning

41715-22 41715-36
CAAA41715-22EA 72.05 CAD
CAAA41715-22 CAAA41715-36
Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate
Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate

CAS No.: 123409-01-8 Molecular Formula: C10H5Na3O9S3.xH2O Formula Weight: 434.31(anhy) Physical Form: Powder MDL No.: MFCD00149263

Formula: C₁₀H₅Na₃O₉S₃.xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149263
CAS Number: 123409-01-8
EINECS: 225-962-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR