Styrene-4-sulfonic acid sodium salt
Supplier: Thermo Scientific Chemicals
41690-18 41690-30
CAAA41690-18EA 89.27 CAD
CAAA41690-18 CAAA41690-30
Styrene-4-sulfonic acid sodium salt
CAS No.: 2695-37-6 Molecular Formula: C8H7NaO3S Formula Weight: 206.19 Physical Form: Powder Appearance: White Beilstein Reference: 3575931 MDL No.: MFCD00013379