Cobalt titanate (<0,15% Ni) ≥99.8%

Supplier: Thermo Scientific Chemicals
Danger

41670-18 41670-30
CAAA41670-18EA 214.88 CAD
CAAA41670-18 CAAA41670-30
Cobalt titanate (<0,15% Ni) ≥99.8%
Cobalt titanate

Purity: 99.8% CAS No.: 12017-01-5 Molecular Formula: CoTiO3, may contain small amounts of other phases Formula Weight: 154.83 Physical Form: -325 Mesh Powder MDL No.: MFCD00016022

Formula: CoO₃Ti
MW: 154.797 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016022
CAS Number: 12017-01-5
EINECS: 234-615-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR