Bismuth titanium oxide ≥99.9%

Supplier: Thermo Scientific Chemicals

41669-18 41669-30
CAAA41669-18EA 181.7 CAD
CAAA41669-18 CAAA41669-30
Bismuth titanium oxide ≥99.9%
Bismuth titanium oxide

Purity: 99.9% CAS No.: 12233-34-0 Molecular Formula: Bi2Ti4O11 Formula Weight: 785.42 Physical Form: -325 Mesh Powder Odor: Odorless MDL No.: MFCD01310393

Formula: Bi₁₂TiO₂₀
MW: 785.42 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01310393
CAS Number: 12441-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR