Ditungsten pentaboride ≥99.4% (metals basis)

Supplier: Thermo Scientific Chemicals

41664-14 41664-22
CAAA41664-14EA 106.49 CAD
CAAA41664-14 CAAA41664-22
Ditungsten pentaboride ≥99.4% (metals basis)
Ditungsten pentaboride

Purity: 99.4% CAS No.: 12007-98-6 Molecular Formula: W2B5, may contain small amounts of other phases Formula Weight: 421.76 Physical Form: -325 Mesh Powder Odor: Odorless MDL No.: MFCD00049707

Formula: B₅W₂
Storage Temperature: Ambient
MDL Number: MFCD00049707
CAS Number: 12007-98-6
EINECS: 234-524-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR