Polyvinylpyrrolidone cross-linked

Supplier: Thermo Scientific Chemicals

Synonyms: Polyvinylpolypyrrolidone, N-Vinyl-2-Pyrrolidone

41631-22 41631-36
CAAA41631-22EA 88.01 CAD
CAAA41631-22 CAAA41631-36
Polyvinylpyrrolidone cross-linked
Polyvinylpyrrolidone cross-linked

CAS No.: 25249-54-1 Molecular Formula: (C6H9NO)n Physical Form: Powder Appearance: White Odor: Odorless Melting Point: 300° MDL No.: MFCD00149016

Formula: C₆H₉NO
MW: 111.144 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149016
CAS Number: 25249-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR