Copper(II) methacrylate, tech.

Supplier: Thermo Scientific Chemicals
Warning

41621-06 41621-14
CAAA41621-06EA 95.59 CAD
CAAA41621-06 CAAA41621-14
Copper(II) methacrylate, tech.
Copper(II) methacrylate

CAS No.: 19662-59-0 Molecular Formula: C8H10CuO4 Formula Weight: 233.71 Physical Form: Powder Appearance: Dark blue-green MDL No.: MFCD00080547

Formula: C₈H₁₀CuO₄
Storage Temperature: Ambient
MDL Number: MFCD00080547
CAS Number: 19662-59-0
EINECS: 243-211-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR