Hafnium(IV) ethanolate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hafnium ethoxide, Hafnium(IV) ethoxide
41607-06
41607-14
CAAA41607-06EA
287.56
CAD
CAAA41607-06
CAAA41607-14
Hafnium(IV) ethanolate ≥99.9% (metals basis)
Hafnium (IV) ethanolate
Preparation of refractory coatings
Purity: 99.9%
CAS No.: 13428-80-3
Molecular Formula: Hf(OC2H5)4
Formula Weight: 358.73
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Boiling Point: 180°-200°/13mm
UN No.: 3180
MDL No.: MFCD00070459
Preparation of refractory coatings
Formula:
C₈H₂₀HfO₄ Boiling Pt: 180…200 °C (13 mmHg) Melting Pt: 178 °C…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00070459 CAS Number: 13428-80-3 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...