Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.1 mm (0.004 in) Type 304
Supplier: Thermo Scientific Chemicals
|
Danger
|
41583-FF
41583-FI
41583-GH
CAAA41583-FFEA
47.56
CAD
CAAA41583-FF
CAAA41583-FI
CAAA41583-GH
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.1 mm (0.004 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Odor: Odorless
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...