Polyethylene glycol monomethyl ether, M.W. 550
Supplier: Thermo Scientific Chemicals
41561-22
41561-36
CAAA41561-22EA
58.46
CAD
CAAA41561-22
CAAA41561-36
Polyethylene glycol monomethyl ether, M.W. 550
Polyethylene glycol monomethyl ether
CAS No.: 9004-74-4
Molecular Formula: CH3(OCH2CH2)nOH
Flash Point:
Formula:
CH₃(OCH₂CH₂)nOH Melting Pt: 18…20 °C Density: 1,10 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...